Learn More
ACROBiosystems ACROBiosystems Human IL-6 R alpha / CD126 Protein, Fc Tag
Catalog No. 501957081
Encompass_Preferred
Recombinant Protein;1MG;Human IL-6 R alpha, Fc Tag (ILR-H5259) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Pro 365 (Accession # P08887-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 65.0 kDa. The protein migrates as 70-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-6, Tag Free (Cat. No. IL6-H4218) at 5 μg/mL (100 μL/well) can bind Human IL-6 R alpha, Fc Tag (Cat. No. ILR-H5259) with a linear range of 1-20 ng/mL (QC tested).
Catalog No. 50-195-7081 Supplier ACROBiosystems Supplier No. ILRH52591MG
Recombinant Protein;1MG;Human IL-6 R alpha, Fc Tag (ILR-H5259) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Pro 365 (Accession # P08887-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 65.0 kDa. The protein migrates as 70-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-6, Tag Free (Cat. No. IL6-H4218) at 5 μg/mL (100 μL/well) can bind Human IL-6 R alpha, Fc Tag (Cat. No. ILR-H5259) with a linear range of 1-20 ng/mL (QC tested).