Learn More
Medchemexpress LLC HY-P7082 10ug Medchemexpress, IL-7 (CHO-expressed), Mouse CAS: Purity:>98%
SDP
Catalog No. 501934322
Encompass_Preferred
Medchemexpress, HY-P7082 10ug IL-7 (CHO-expressed), Mouse CAS: Recombinant Mouse Interleukin-7 (CHO-expressed) is constitutively produced by stromal cells from the bone marrow and thymus, plays a crucial role in B cell lymphopoiesis and in T cell homeostasis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4322 Supplier Medchemexpress LLC Supplier No. HYP708210UG
Medchemexpress, HY-P7082 10ug IL-7 (CHO-expressed), Mouse CAS: Recombinant Mouse Interleukin-7 (CHO-expressed) is constitutively produced by stromal cells from the bone marrow and thymus, plays a crucial role in B cell lymphopoiesis and in T cell homeostasis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.