Learn More
Sino Biological Recombinant Human IL-7 Protein, HPLC-verified
Supplier:  Sino Biological 11821HNAE100
Encompass_Preferred
A DNA sequence encoding the mature form of human IL7 (NP_000871.1)(Asp26-His177) was expressed with a N-terminal Met.
Catalog No. 50-211-6280