Learn More
CPC Scientific H-Ala-Val-Leu-Pro-Arg-Ser-Ala-Lys-Glu-Leu-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific ITLK001A
Encompass
The -5 to +5 N-terminal fragment of the 77 amino acid form of IL-8 exhibited potent chemotactic acitivity in human neutrophils with an EC50 value of 2 nM, but it did not bind to the IL-8 receptors.ONE-LETTER SEQUENCE: AVLPRSAKELMOLECULAR FORMULA: C48H86N14O14MOLECULAR WEIGHT:1083.3STORAGE CONDITIONS: -20 5CRESEARCH AREA: ImmunomodulationREFERENCES: S.K.Thompson et al., Int. J. Pept. Protein Res., 47, 214 (1996)
Catalog No. 50-210-9610