Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-8 (CXCL8) Canine ProcartaPlex™ Simplex Kit

Catalog No. 502557073
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-255-7073 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-255-7073 Supplier Invitrogen™ Supplier No. EPX01A50506901
Add to Cart
Add to Cart

The Canine IL-8 (CXCL8) Simplex ProcartaPlex Kit measures IL-8 (CXCL8) protein and is designed to be combinable with other Simplex kits so that you can create your own multiplex panel that utilizes Luminex xMAP technology for protein detection/quantitation.

The Canine IL-8 (CXCL8) Simplex ProcartaPlex Kit measures IL-8 (CXCL8) protein and is designed to be combinable with other Simplex kits so that you can create your own multiplex panel that utilizes Luminex xMAP technology for protein detection/quantitation. When combining multiple Simplex kits (i.e., when you are not using a pre-configured Multiplex Panel), only one buffer kit (sold separately) is needed for each assay plate regardless of plex size.

About ProcartaPlex Assays for the Luminex Platform

ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.

  • More results per sample—measure multiple protein targets simultaneously in a single 25–50 μL sample
  • Well-established Luminex technology—highly referenced multiplexing platform for protein detection and quantitation

ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 80 protein targets in a single 25–50 μL sample from plasma, serum, cell culture supernatants, and other bodily fluids.

The Luminex beads in the ProcartaPlex assay are internally dyed with precise proportions of red and infrared fluorophores to create spectrally unique signatures that can be identified by the Luminex xMAP detection systems (e.g. Luminex 200, FLEXMAP 3D, and MAGPIX). Similar to a sandwich ELISA, the ProcartaPlex assay uses matched antibody pairs to identify the protein of interest. In a multiplexed assay, each spectrally unique bead is labeled with antibodies specific for a single target protein, and bound proteins are identified with biotinylated antibodies and streptavidin–R-phycoerythrin (RPE). The conjugation of protein-specific antibodies to a distinct bead allows for analysis of multiple targets in a single well.

The most significant difference between a ProcartaPlex assay and ELISA is that the capture antibody in the ProcartaPlex assay is conjugated to a bead and not adsorbed to the microplate well, so the ProcartaPlex assay reagents are free-floating in the solution. For detection, the Luminex 200 instrument, for example, contains two lasers, one to distinguish the spectral signature of each bead and the second to quantify the amount of RPE fluorescence, which is proportional to the amount of protein present in the sample. ProcartaPlex multiplex assays can profile more target proteins using significantly less sample in the same time that it takes to perform a traditional sandwich ELISA.

ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, nonhuman primate, porcine, and canine). Visit thermofisher.com/procartaplex for more information, including a comprehensive list of individual protein targets.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Simplex Kit
Quantity 96 Tests
Species Canine
Combinability Combinable
Bead Type IL-8 (CXCL8) [27]
Assay Sensitivity As determined for Lot 1: 6.9 pg/mL
Assay Range As determined for Lot 1: 10.08 - 41,300 pg/mL
Format Simplex Kit
For Use With (Equipment) Luminex™ Instruments
Gene C-X-C motif chemokine ligand 8
Gene Alias IL8
Gene ID (Entrez) 403850
Gene Symbol CXCL8
Product Line ProcartaPlex
Protein Interleukin-8
Protein Subtype IL-8, C-X-C motif chemokine 8, Chemokine (C-X-C motif) ligand 8
Sample Type Serum, Plasma, Cell Culture Supernatants
Sample Volume Serum, Plasma: 25 μL, CCS: 50 μL
UniProt ID P41324
Content And Storage

• 1 vial Capture Beads (50X)
• 1 vial biotinylated Detection Antibody (50X)
• 2 vials Canine Standard Mix Cytokine/Chemokine/Growth Factor (lyophilized)

Store at 2°C to 8°C.

Shipping Condition Wet Ice
Research Area Immunology, Oncology, Neurobiology, Toxicology
Show More Show Less
What is the size of the Luminex beads you currently use?

The beads used in our Luminex instrument-compatible ProcartaPlex and QuantiGene Plex assays are 6.5 micron superparamagnetic beads.

I am interested in performing Luminex assays using BioSource kits, and I have a Luminex xMAP system. Besides the kits and system, what other reagents and equipment will I need?

The following is a list of general lab supplies that are required for running BioSource immunoassays on the Luminex xMAP system:
1) Sonicating water bath
2) Orbital shaker
3) Vortexer
4) Repeating and/or multi-channel pipetter (not required, but recommended)
5) Calibrated adjustable precision pipettes, with disposable plastic tips
6) Glass/plastic tubes and racks for preparing reagents
7) Graduated cylinder and container for preparing wash solution
8) Aluminum foil
9) Deionized or distilled water.

Do the Luminex beads require special care in handling?

The Luminex beads should be protected from light because they are susceptible to photobleaching. We recommend protecting the beads by keeping containers covered with aluminum foil during all incubation steps, and exercising care during handling. The beads should not be frozen, subjected to excessive heat, or exposed to organic solvents.

Why would the Luminex acquisition software display "Sample Empty" messages during analysis?

(1) The user did not properly aliquot the diluted beads, such that no beads were actually added to the wells (make sure that the bead concentrates are sonicated and vortexed well, then check the pipet tip to ensure that air bubbles were not drawn up)
(2) The user missed loading diluted beads to some wells, which is likely since the small volume is clear and difficult to visualize in the clear plastic plate (we have now addressed this customer difficulty by coloring each of the Buffer Reagent Kit components)
(3) The user applied too much vacuum pressure at some point during the wash steps, or allowed the pressure to spike even once, such that the filter membrane tore in a few wells releasing the beads (make sure that the vacuum manifold pressure is kept below 5mm/in Hg, depending on their system -- a good rule of thumb is that it should take a full 3-second count to GENTLY empty the wells of 200uL)
(4) The user did not properly sonicate and vortex the beads prior to dilution, such that the percent of bead aggregation was high and the instrument was unable to find enough single beads to meet the events/bead value designated by the customer (make sure that the Bead Concentrate tube is put into the waterbath all the way to the cap, since the tube is hollow until the top third)
(5) The user lost beads by shaking the plate too aggressively or handling it improperly (make sure that the orbital shaker is set to a speed that allows for maximum vortex in the wells without spillage)
(6) The user exposed the beads to an excess of light during storage or running of the assay, such that some but not all of the beads were photobleached and therefore falling outside the acceptable range for each bead region (make sure that the plate is covered on the top/sides with foil throughout the assay, away from Windows and spotlights, and that the bead component of the kits is stored in the dark)*
(7) There was a clog in the sample needle, such that the instrument was unable to take up enough sample to meet the number of events requested per bead region (suggest that the user follow the manual instructions for dislodging a clog, which include several Back Flush steps and may require removal of the needle for sonication with probe alignment).

* Some of the older Antibody Bead Kits still have clear plastic tops instead of black ones. In cases where customers store kits in lit refrigerators, or keep them open on the lab bench, even a few hours of light exposure is enough to photobleach beads. It is important to note, in general, that higher number bead regions are more susceptible to photobleaching. In order to draw conclusions about the source of the difficulty, we would ask to see the data, specifically the Masterplex QT file, which would enable us to examine the pattern of "Sample Empty" occurrences in addition to the bead counts per well.

What are the Luminex beads made of?

The beads are made of polystyrene.

How do the Luminex 200 Instrument System, Luminex FLEXMAP3D Instrument System and Luminex xMAP INTELLIFLEX System simultaneously monitor concentrations of several different analytes in a single sample?

Each protein detected requires a different bead region to differentiate between analytes. The Luminex 200 Instrument System, Luminex FLEXMAP3D Instrument System and Luminex xMAP INTELLIFLEX System are outfitted with two lasers. The first laser is a red diode laser that classifies beads (region) based on color and hence identifies which protein is being measured. The second laser is a green YAG laser that quantitates reporter fluorophore (R-Phycoerythin, RPE) associated with the bead surface and determines the amount of the protein that is bound to the bead. The beads pass through the system at a rate of several thousand per second, excited first by the red laser and then the green laser. The results obtained show protein identity and the intensity of the signal, respectively. The intensity of the signal is directly proportional to the concentration of the analyte captured in the sample. Therefore, by monitoring the spectral properties of the beads and the amount of associated RPE fluorescence, the concentration of one or more proteins can be determined.

Do you offer a Mac-compatible version of the ProcartaPlex Analyst Software?

Unfortunately, we do not currently offer a Mac-compatible version of the ProcartaPlex Analyst Software. ProcartaPlex Analyst Software is compatible with PC, or xPONENT Software is available on Luminex instruments to analyze your data.

Which Luminex instruments are available for multiplex detection and analysis?

Here (https://www.thermofisher.com/us/en/home/life-science/antibodies/immunoassays/procartaplex-assays-luminex/luminex-instruments.html) is an overview of Luminex instruments we offer for multiplex detection and analysis.

I am getting poor recovery with my ProcartaPlex multiplex immunoassay. What can I do?

Here are possible causes and solutions:
- The wrong cell culture media may have been used to prepare the standards. We recommend using the same cell culture media that was used to culture the cells.
- The samples and antigen standards may not have been stored on ice. We recommend preparing the samples and standards on ice before setting up the assay.

I am seeing low signal or sensitivity in my ProcartaPlex multiplex immunoassay. What should I do?

Standards may not have been reconstituted and diluted correctly. We recommend preparing fresh antigen standards following the instructions provided in the corresponding ProcartaPlex user guide.

With my ProcartaPlex multiplex immunoassay, I am getting a low bead count. What is causing this?

Here are possible causes and solutions:
- Volume of the bead solution is too low. We recommend adding 120 µL Reading Buffer into each well and shaking at 600 rpm for 5 mins at room temperature to resuspend beads before reading on the Luminex instrument.
- High bead aggregation. We recommend vortexing the bead suspension well before using in the assay and ensuring that the beads are properly mixed during the incubation steps.
- Dyes contained in the beads are photo-bleached from overexposure to light. We recommend storing the bead solution and the 96-well plate in the dark.
- Samples causing the instrument to clog. We recommend removing the 96-well Flat Bottom Plate and performing a wash and rinse to the instrument and then re-running the assay with further dilution of samples.
- Probe height is incorrect. We recommend referring to the Luminex manual for proper adjustment of the needle height.
- The instrument needle is partially clogged. We recommend replacing or cleaning the needle according to the manufacturer's recommendations.
- Beads stuck to the bottom of the plate. We recommend confirming that the plate shaker is set to 600 rpm and shaking for at least 5 mins before reading.
- Air bubble in the sample loop. We recommend referring to the specific Luminex manual for your assay for proper removal of the air bubble.

In ProcartaPlex multiplex immunoassays, we are seeing a limited dynamic range for BioPlex software users. Why is this?

The instrument may have been calibrated at high PMT settings. We recommend calibrating the instrument using the CAL2 Low RP1 target value.

I am seeing high CVs with my ProcartaPlex multiplex immunoassay. Why is this?

Here are possible causes and solutions:
- Samples and antigen standards not stored on ice. We recommend preparing the samples and standards on ice before setting up the assay.
- Contamination from re-using the Plate Seal. We recommend using a new Plate Seal for each incubation step.
- Incomplete washing. After adding the standards and samples, it is very important that any excess standards are removed during the wash step.
- Contamination from contents from adjacent wells. Avoid splashing the Wash Buffer during wash steps into adjacent wells.
- Poor pipetting techniques. We recommend using a multichannel pipettor and careful pipette techniques. Avoid touching pipette tips to sides of the wells when adding Wash Buffer.

With my ProcartaPlex multiplex immunoassay, I am getting a low flow rate. What should I do?

Samples/beads could have been stuck in the flow cell. We recommend removing the 96-well plate and performing a wash and rinse cycle.

Do you offer FlowCytomix assays?

Sorry, the FlowCytomix product line has been discontinued. However, we offer a new antibody and magnetic bead-based detection system, i.e., ProcartaPlex assays for multiplex protein quantitation using the Luminex instrument platform.

How long does it take to acquire ProcartaPlex assay data on the different Luminex instruments?

With everything working smoothly, here is the approximate read time for one plate on different Luminex instruments:

- FLEXMAP 3D Systems and INTELLIFLEX Systems will read 96 well plates in ~20 min, and 384 well plates in ~75 min.
- Luminex 100/200 Systems will read 96 well plates in ~40 min.
- MAGPIX System will read 96 well plates in ~50-60 min. Please note that these are estimates for the read times of the plates only, and do not include time for calibration or set-up of the instrument.

Is the sensitivity value for a Multiplex ProcartaPlex assay different when compared to the sensitivity for those same analytes assayed as Simplex assays?

In general, sensitivity between Simplex and Multiplex ProcartaPlex assay kits is comparable.

Why does the ProcartaPlex ULOQ (Upper Limit of Quantitation) and LLOQ (Lower Limit of Quantitation) change between different lots of standards?

We supply the standard protein in a lyophilized form. Lyophilization is a dehydration process that preserves the perishable standard. We have demonstrated that often, the amount of starting protein prior to lyophilization and that of the final product after lyophilization varies (this could be different from analyte to analyte). To ensure that we deliver the most quantitative and reproducible product, we always calibrate the standard after lyophilization and therefore, the concentration range can vary from lot to lot of the standard.

Can the ProcartaPlex beads be frozen?

No. Storage of the beads at temperatures below 0 degrees C will damage the beads and render them unusable.

What is the minimum bead count used for statistical significance/analysis in ProcartaPlex assays?

We validate and release our ProcartaPlex assays with a minimum bead count of 50. In our experience, the assays also work with a bead count of 20 though we have not validated them at this bead count.

Is ProcartaPlex assay sensitivity information based on data using the 2 hr or overnight incubation with samples?

The sensitivity information is based on the 2 hr incubation with samples.

How can I minimize inter-assay variation for ProcartaPlex assays?

To minimize inter-assay coefficient of variation between two independent assay plates, it is very important that the same operator performs the assays. Also, we recommend that you follow the incubation times exactly as they are stated in the manual in order to get consistent results. Other tips for minimizing inter-assay coefficient of variation include performing a standard curve in duplicate on each plate and using the same kit lot.

I am trying to find out whether the medium/serum used to culture my cells will interfere with the ProcartaPlex assay. Which cell culture media have you tested in-house with ProcartaPlex assays?

In-house, we test ProcartaPlex assays with RPMI + 0.5% FCS. You would have to empirically determine if your specific medium will interfere with the assay.

Note: Our customers have successfully used media containing up to 5% FBS with ProcartaPlex assays. As bovine serum contains TGF beta, the use of FBS or FCS should be avoided with TGF beta ProcartaPlex assays.

I am a Bio-Plex user. How can I generate and export CSV raw data to be analyzed using the ProcartaPlex Analysis App?

Open the Bio-Plex Manager and click on “File”, then “Document Export Properties”, then “Output CSV Format” and set “Analytes Labels” to “Name (Region)”. Choose the desired file “Destination”, and click “OK”. Click “Document Export”, then “Export”, and import the generated .CSV file into ProcartaPlex Analysis App on Thermo Fisher Connect: https://apps.thermofisher.com/apps/procartaplex

How do I process tear samples for use in a Luminex assay?

We have not specifically tested ProcartaPlex assays with tear samples in-house. However, here is a reference from the literature describing the use of ProcartaPlex assays with tear samples (https://www.ncbi.nlm.nih.gov/pmc/articles/PMC4161422/pdf/pone.0107370.pdf).

How do I process breast milk samples for use in a Luminex assay?

We have tested breast milk samples in-house using the serum ProcartaPlex protocol and found no general interference with the breast milk matrix. Please note that we have only tested a few analytes (TNFa, IL-10, TNFR), however, there is no obvious reason why other analytes should not work as well.

What is the size of beads used in the ProcartaPlex assays?

The size of the beads in our ProcartaPlex assays is 6.5 µm.

What data analysis software should I use to analyze the data obtained with my ProcartaPlex assay?

All Luminex instruments come with xPonent data analysis software that can be used. Alternatively, we offer a free and robust data analysis software package, that can be downloaded from our website (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/luminex-multiplex-assays/procartaplex-immunoassays/procartaplex-analyst-software.html).

Additionally, please refer to our ProcartaPlex Assays Support Center (https://www.thermofisher.com/us/en/home/technical-resources/technical-reference-library/antibodies-immunoassays-support-center/luminex-assays-support.html) for data analysis tips and tricks.

My ProcartaPlex assay data lists some targets as OOR<, what does this mean?

This means that the data obtained in that run are below the detectable limit of the assay. Here are some possibilities and suggestions:
1. Qualify your standard curve using the data analysis tools found in our Luminex Assays Support Center (https://www.thermofisher.com/us/en/home/technical-resources/technical-reference-library/antibodies-immunoassays-support-center/luminex-assays-support.html).
2. Make sure that your dilution factors are set correctly.
3. It is possible that the functional sensitivity of the standard curve can be extended to better distinguish the data at the lower end of the curve. This can be done by adding one or two further dilutions to the curve and re-running the assay with the samples of lower concentrations.
4. It is possible that the levels of your protein of interest fall below the detection limits of the assay.

Where can I find a complete list of targets for your entire ProtcartaPlex assay portfolio?

A complete list of available targets for our entire ProtcartaPlex assay portfolio can be found on our website by using the ProcartaPlex Panel Configurator (https://www.thermofisher.com/order/luminex/) and choosing the species of interest. Alternatively, this information can be found on our ProcartaPlex Assays page here: https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/luminex-multiplex-assays/procartaplex-immunoassays/procartaplex-assays.html

Where can I find a complete list of targets for your human ProcartaPlex assays?

A complete list of available targets for our entire ProtcartaPlex assay portfolio can be found on our website by using the ProcartaPlex Panel Configurator (https://www.thermofisher.com/order/luminex/) and choosing the species of interest. Alternatively, a list of all human ProcartaPlex kits can be pulled up on our website using the search term 'ProcartaPlex Assay Human', or by opening the drop-down menus for Human ProcartaPlex Panels on our ProcartaPlex Assays page (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/luminex-multiplex-assays/procartaplex-immunoassays/procartaplex-assays.html).

My ProcartaPlex assay custom panel selection says: "TGF-beta1 has to be acid treated for proper detection of bioactive form. Recommendation: single plex assay. Alternative: LAP (TGF-beta1)" What is the difference between TGF-beta1 and LAP (TGF-beta1)?

The TGF-beta1 assay requires an acid pre-treatment of the sample to reveal the TGF-beta1 protein and therefore, the assay cannot be combined with other assays. The acid pre-treatment of the sample will destroy the other protein epitopes. However the LAP TGF-beta1 assay does not require an acid treatment, but it will measure only the LAP-TGFbeta1 complex. Neither purified LAP nor purified TGF-beta1 alone can be measured in this particular assay.

My ProcartaPlex assay custom panel selection has notes regarding different dilution factor needed for some of my targets, can these targets still be combined into one panel?

This will depend on your sample type. Often serum and plasma may require dilutions for certain targets whereas culture supernatant does not. However, it is optimal to combine only those targets that require the same dilution.

My ProcartaPlex assay custom panel selection shows bead region conflicts, can my targets still be combined into one panel?

Yes, our Mix & Match team can adjust the bead regions as necessary to accommodate targets in your panel and accommodate instrument parameters (i.e., MagPix can only read 50 bead regions).

I want to run half a plate at a time for my ProcartaPlex assay, can I use the same plate again? Can I purchase more plates?

Yes, you can use half a plate at a time, but make sure to seal the unused half with plate sealing tape to prevent any contamination during the assay. Alternatively, you can purchase extra plates (Cat. No. EPX-88182-000).

Can I purchase more of the Standard Mix for my ProcartaPlex assay?

This may be possible as a custom order. Please reach out to your Sales Representative or to Technical Support (techsupport@thermofisher.com) with your kit number and lot number, they can work with our manufacturing team to determine availability of lot-specific standards.

Which hand-held magnetic separator should I use with your ProcartaPlex assay kits?

Our ProcartaPlex assays have been validated with the Hand-Held Magnetic Plate Washer (Cat. No. EPX-55555-000). We also recommend the Magnetic 96-Well Separator (Cat. No. A14179) as it has a stable magnetic field distributing beads along the bottom of the wells for maximum retention of beads during wash steps.

Can I use automated plate washers with your ProcartaPlex assays?

If you have a magnetic automated plate washer, this can be used with our magnetic bead kits.

I need more ProcartaPlex Streptavidin-PE for my assay. Can I purchase this item separately?

Yes, ProcartaPlex Streptavidin-PE (Cat. No. EPX-SAPE-000) is available as a stand-alone item. We offer most of our PorcartaPlex buffers and reagents as stand-alone items. A complete list of what is currently available can be found on our website here: https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/luminex-multiplex-assays/procartaplex-immunoassays/procartaplex-supplemental-products.html.

I have spilled my ProcartaPlex assay buffer, can I purchase more?

Yes, our Universal Assay Buffer (Cat. No. EPX-11110-000) can be bought separately. We offer most of our ProcartaPlex buffers and reagents as stand-alone items. A complete list of what is currently available can be found on our website here, under "Accessories": https://www.thermofisher.com/us/en/home/life-science/antibodies/immunoassays/procartaplex-assays-luminex/procartaplex-immunoassays/procartaplex-preconfigured-panels.html.

How long does a typical ProcartaPlex assay take to set up and run?

Typical timeline for sample to results for our ProcartaPlex assays is 4.5 hrs.

Do you have any references for your ProcartaPlex assays?

We do list many references for our ProcartaPlex portfolio on our website here: https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/luminex-multiplex-assays/procartaplex-immunoassays/procartaplex-immunoassay-publications.html. More recent publications can be found using internet search tools such as Google Scholar or similar.

What species of non-human primates can be used with your NHP ProcartaPlex assays?

We have listed all the species we have tested in our Cross-reactivity Chart for NHP ProcartaPlex Simplex Kits. This chart can be found here (https://www.thermofisher.com/us/en/home/life-science/protein-biology/protein-assays-analysis/luminex-multiplex-assays/procartaplex-immunoassays/procartaplex-assays.html) by clicking on the link for Non-human Primate ProcartaPlex Panels.

What is a ProcartaPlex High Sensitivity assay?

ProcartaPlex High Sensitivity assays are designed to measure small concentration differences in cell culture supernatants, plasma, and serum samples with 10-fold lower LLOQs (Lower Limit of Quantification) and sensitivities for all analytes in the femtogram range. They have been further optimized to overcome the sensitivity detection limits of conventional multiplex immunoassays.

What is the difference between the "Convenience" and the "Combinable" multiplex panels? Can they be multiplexed with each other?

“Convenience” multiplex panels are pre-mixed and optimized bead sets for use without modification. “Combinable” multiplex panels are target sets which can be combined or supplemented by addition of beads from ProcartaPlex Simplex kits. These two types of panels cannot be multiplexed together.

How do I set up the Luminex instrument?

Please refer to the assay protocol for kit-specific instructions and your Luminex instrument user guide for instrument-specific instructions. Prior to running the assay, please ensure that the probe height has been calibrated with the 96-Well Flat Bottom Plate supplied with the kit. Failure to adjust the probe height can cause damage to the instrument or low bead count. The Luminex system allows for calibration of low and high RP1 target values. We recommend RP1 low target value settings for immunoassays. Please refer to the lot-specific Certificate of Analysis provided with the kit for bead region and analyte associations when entering the information into the Luminex Acquisition Software.

I am planning to run tissue lysates for use on a Procartaplex assay. Can you please provide the protocol?

Please follow the instructions here (https://tools.thermofisher.com/content/sfs/manuals/Preparation-of-Tissue-Homogenate.pdf).

My samples have been fixed with paraformaldehyde and processed to separate the serum/plasma fraction after fixation. Are they compatible with ProcartaPlex assays and how should I process them?

We have not tested the use of fixed samples with the ProcartaPlex assay and therefore cannot confirm compatibility at this time.

I have bought several ProcartaPlex Simplex Bead Sets. Which premixed standard do I have to use in my assay?

Please refer to the lot-specific Certificate of Analysis to check the source of the Standard Mix (i.e., Standard Mix A, B, or C) included with each kit. When combining Simplex Bead Sets with other Simplex Bead Sets or Multiplex kits, multiple vials of the same premixed standard (Standard Mix A, B, or C) may be shipped. If the combination of analytes you want to analyze in your multiplex assay includes two analytes that require the same premixed standard, use only one vial of the premixed standards to prepare the standard curve. If the kits you want to combine include different lots of the same premixed standard, please choose one vial (of any lot) for your multiplex assay.

How many standard points do we need to prepare when using the ProcartaPlex kits?

You will need to prepare a 7-point standard curve as well as a blank.

Can a finished ProcartaPlex plate be read multiple times on the Luminex instrument without a significant signal loss?

Yes, it is possible to reread a finished ProcartaPlex plate without a loss in the signal or the number of beads counted. Please note that the Luminex instrument adds additional liquid to the wells with each analysis. It is possible that the wells may become overfilled with fluid after the third analysis. Hence, we do not recommend reading the ProcartaPlex plates more than two times.

Can I combine ProcartaPlex kits, which were not purchased on the same order? For example: I purchased a kit 3 months ago and another kit today. Can I combine them or do the kits have to be shipped on the same order?

Yes, you can combine ProcartaPlex Combinable panels and Simplex kits from different orders to run a bigger multiplex experiment. You will want to ensure that there is no overlap of bead regions between the kits you are combining. Please refer to the online ProcartaPlex Panel Configurator (https://www.thermofisher.com/order/luminex).

Can I use the ProcartaPlex Mouse Basic Kit with ProcartaPlex Human Simplex and vice versa?

No, the Basic Kits are specific to the species listed, i.e. the Mouse Basic Kit can only be used with Mouse Simplex Bead Sets. The Mouse, Human, Canine, Porcine, Rat and NHP ProcartaPlex Basic Kits are not interchangeable.

Do I need to purchase a Basic Kit when combining Simplex kits with a Multiplex ProcartaPlex immunoassay kit?

When combining a Multiplex kit with Simplex Bead Sets, a Basic kit is not required. All the non-target specific reagents which are included in the Basic kits are components of our Multiplex kits. Only when you are combining multiple Simplex kits (i.e., no Multiplex kit in the panel) will you need to also purchase a Basic kit.

Is it possible to measure ProcartaPlex Luminex beads on a flow cytometer?

No. To analyze ProcartaPlex (Luminex) plates, you will need access to a Luminex analyzer such as a Luminex 100/200, MAGPIX, or FLEXMAP 3D system.

I have completed the ProcartaPlex multiplex assay but can't analyze the assay plate immediately. How long can I store the ProcartaPlex plate before analysis on a Luminex instrument without a significant loss of signal?

Ideally, we recommend that you analyze the samples immediately. If the plate cannot be read on the day of the assay, cover and store the plate in the dark, overnight at 2-8 degrees C for reading the following day without significant loss of fluorescence intensity. When you are ready to read the plate, bring the plate to room temperature on an orbital plate shaker (500-600 rpm), still protected from light. Perform a wash step using 100 µL of fresh, room temperature Working Wash Solution/Reading Buffer and perform the analysis. We do not recommend storing the ProcartaPlex assay plate longer than 1 day.

During my ProcartaPlex assay data analysis, I am getting a warning message that there is high bead aggregation. What should I do?

Here are possible causes and solutions for this issue:

- Check the protocol settings (make sure you select the correct DD settings).
- Check the level of sheath fluid and empty the waste.
- Before acquiring the plate, run calibration and verification beads on the Luminex instrument.

During my ProcartaPlex assay data analysis, the beads fall below or to the lower left of their bead region on the bead map. Why is this?

Here are possible causes and solutions for this issue:

This usually indicates that the beads have been photobleached. This problem can also be caused by exposing the beads to organic solvents. Unfortunately, the assay will have to be repeated because the beads cannot be restored. The beads must be protected from light and organic solvents.
Alternatively, the instrument may be off in its measurements or you may have a calibration issue. Call the manufacturer for a service appointment.

During my ProcartaPlex assay data analysis, beads do not appear in the region gated. What happened?

This indicates that an incorrect buffer was used for the final step. The Wash Solution provided in the kit must be used for washing the beads and the Reading Buffer should be used for resuspending the beads before loading them into the Luminex instrument. The osmolarity of the solution will impact the size of the bead, and any change in the bead size will alter detection by the instrument.

The bead counts for all of my ProcartaPlex assay wells are erratic. What went wrong?

Here are some suggestions:

- Before acquiring the plate, run calibration and verification beads on the Luminex instrument.
- Review the instrument settings and make sure they are appropriate for the assay being run (adjustment of needle height, make sure you select the correct bead gates and the correct DD settings).
- Shake the plate before acquisition on the instrument to resuspend the beads.
- Vortex the beads for 30 sec before adding them into the plate.
- Washing: Do not forget to keep the plate for about 2 mins on the Hand-Held Magnetic Plate Washer before emptying the plate.

The bead count for my ProcartaPlex assay standard curve is correct and regular. The bead count for my samples is erratic and I get a warning message saying "The acquisition had at least one region that did not reach the maximum count". What should I do?

This pattern is indicative of a sample matrix effect. Here are some suggestions:

- Confirm that the sample has been clarified and is free of debris and free of lipids (5-10 min centrifugation recommended).
- Confirm that there is at least a 1:1 ratio of sample to assay diluent for serum, plasma samples. For cell lysates or tissue homogenates, confirm that the sample has been diluted appropriately in assay buffer to reduce the concentration of detergent in the lysis buffer to ⋜0.01%. For other sample types, further sample optimization may be required.

The readout for my ProcartaPlex assay samples says that they are below the detectable limit. What are your suggestions?

Here are some suggestions:

- It is possible that the levels of your protein of interest fall below the detection limits of the assay. High Sensitivity Multiplex kits are available for most cytokines.
- Qualify your standard curve (look for plateaus, abnormal curve fits, outliers).

The readout for my ProcartaPlex assay samples says that they are above the detectable limit. What should I do?

Here are some suggestions:

- Sample optimization may be needed: Dilute the sample with an appropriate diluent and re-run.
- Qualify your standard curve (look for plateaus, abnormal curve fits, outliers).

How do I process cell lysates (extract cellular proteins) for the Luminex assay platform?

To process cell lysates (extract cellular proteins), follow the instructions provided here (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0017835_PrepareCellLysates_Procartaplex_PI.pdf).

How do I process saliva samples for the Luminex assay platform?

We have not specifically tested saliva samples in-house and therefore these instructions are only our recommendation. Saliva contains several proteolytic enzymes. It would be important to centrifuge samples and be sure not to pipet any cellular material or debris into the assay plate. We would suggest adding some anti-protease in the sample (for example, trasylol or aprotinine, 10 to 50 U/mL) to protect the protein from enzyme degradation. You may then centrifuge the samples at 1000 x g at 4 degrees C for 10 mins to remove particulates. Use immediately or store aliquots at -80 degrees C. Avoid multiple freeze-thaw cycles.

How do I process oral mucosal transudate for Luminex assays?

We have not specifically tested oral mucosal transudate samples in-house and therefore these instructions are only our recommendation. Isolate the site around the tooth and insert a piece of periodontal filter paper into the gum pocket around the tooth for 30 seconds. Remove the filter paper and extract 4 times with 50 µL PBS for 5 min each at room temperature. The individual extractions can be combined and analyzed. Dilute 2-fold with Assay Diluent before applying to the assay.

How do I process cervical secretion samples for use in a Luminex assay?

We have not specifically tested cervical secretion samples in-house and therefore these instructions are only our recommendation. Cervical sponges should be placed on ice immediately upon collection. Samples should be stored at -20 degrees C for up to one week and then stored at -80 degrees C until ready for assay. After thawing, sponges should be weighed and placed into Eppendorf tubes, using forceps cleaned with ethanol after each transfer. Add 200 µL of ice-cold extraction buffer (recipe below) to each tube and incubate overnight at 4 degrees C. The sponges and extraction buffer can then be transferred to microcentrifuge tubes with 0.2 µm cellulose acetate filters and centrifuged at 13,000 rpm for 10 min at 4 degrees C. The eluate can then be tested for cytokine expression.

Extraction Buffer
50 mM HEPES, pH 7.5
1 mM Na3VO4
150 mM NaCl
1 mM NaF
1 mM EDTA
0.1% Tween 20
25 mM EGTA
10% glycerol

How do I process bronchoalveolar lavage (BAL) for use on the Luminex assay platform?

We have not specifically tested bronchoalveolar lavage (BAL) samples in-house and therefore these instructions are only our recommendation. The bronchoalveolar lavage (BAL) should be collected in a sterile syringe and kept on ice until you are ready to analyze it. Alternatively, BAL can be aliquoted and frozen in usable sample sizes (such that exposure to freeze-thaw is limited to one time). All samples need to be clarified by centrifugation (14,000 rpm for 10 min) and/or filtered prior to analysis to prevent clogging of the filter plates. Dilute 2-fold with Assay Diluent before applying to the plate.

How do I process cerebrospinal fluid (CSF) for use on the Luminex assay platform?

Centrifuge samples at 1,400 rpm for 10 mins at 4 degrees C to remove particulates. Use immediately or store aliquots at -80 degrees C. Avoid multiple freeze-thaw cycles.

How do I process synovial fluid for use on the Luminex assay platform?

We have not specifically tested synovial fluid samples in-house and therefore these instructions are only our recommendation. Collect synovial fluid into non-heparinized tubes and spin at 1,000 x g for 10 min within 30 min of sample collection. The acellular portion of synovial fluid should be stored at -80 degrees C before subsequent analysis. All samples need to be clarified by centrifugation (14,000 rpm for 10 min) and/or filtered prior to analysis to prevent clogging of the filter plates. Dilute samples 1:1 with Assay Diluent prior to addition to the assay. Reference: Raza K et al. (2005) Arthritis Research &Therapy 7(4): R784-R795.

How do I process tissue homogenates for use on the Luminex assay platform?

Collect spleen, lung, brain, kidney, liver, or heart tissue and treat with or without LPS (100 µg, i.p., for 15 mins, 30 mins, 1 hr, 2 hrs, or 3 hrs). Weigh tissue in a 2 mL microcentrifuge tube. Add 500 µL of Cell Lysis Buffer (Cat. No. EPX-99999-000) per 100 mg of tissue. Add one 5-mm Stainless Steel Bead, then assemble tubes into TissueLyser according to the manufacturer’s recommendations. We recommend using 5-mm Stainless Steel Beads from Qiagen (Cat. No. 69989). Homogenize tissue at 25 Hz for 0.5-3 mins as indicated in the table below. Centrifuge the sample at 16,000 × g for 10 mins at 4 degrees C. Transfer the supernatant to a new microcentrifuge tube. Measure the total protein concentration. Dilute samples to 10 mg protein/mL with 1X PBS. To proceed with ProcartaPlex protocol, add 25 µL of Universal Assay Buffer (Cat. No. EPX-11111-000) to 25 µL of the diluted sample to each sample well.

How do I process cell culture supernatants for use on the Luminex assay platform?

Cells should be in log-phase growth. Stimulate cells as desired in appropriate cell culture flasks. Using sterile technique, remove the desired volume of conditioned cell culture medium with a pipette and transfer the medium to clean polypropylene microcentrifuge tubes. Centrifuge the medium at 14,000 rpm for 10 min at 4 degrees C in a refrigerated microcentrifuge to remove any cells or cellular debris. Aliquot the clarified medium into clean polypropylene microcentrifuge tubes. These samples are ready for the assay. Alternatively, clarified medium samples can be aliquoted and stored at -80 degrees C for future analysis. Avoid multiple freeze-thaw cycles. Frozen samples should be allowed to thaw on ice just prior to running the assay. Thawed samples should be clarified by centrifuging at 14,000 rpm for 10 min at 4 degrees C in a refrigerated microcentrifuge prior to analysis to prevent clogging of the Luminex probe and/or filter plate. Follow the assay procedure provided with the kit for appropriate dilutions.

How do I process plasma samples for use on the Luminex assay platform?

Separate the cells from the plasma samples by centrifugation at 2,000 x g for 10 min in a refrigerated centrifuge. Centrifugation at this force is necessary to deplete the sample of platelets. Transfer the supernatant to a chilled clean polypropylene tube with a sterile Pasteur pipette. Maintain the samples at 2-8 degrees C while handling.

If the plasma is to be analyzed at a later date, apportion it into aliquots in polypropylene microcentrifuge tubes and store at -80 degrees C. Avoid multiple freeze-thaw cycles. When you are ready to analyze them, allow the samples to thaw on ice. All plasma samples should be clarified by centrifugation (14,000 rpm for 10 min at 4 degrees C) in a refrigerated microcentrifuge immediately prior to analysis. Follow the assay procedure provided with the kit for appropriate dilutions.

How do I process serum samples for use on the Luminex assay platform?

Serum samples should be collected in pyrogen/endotoxin-free tubes. Whole blood should be allowed to clot for 20-30 mins at 20-25°C. Centrifuge at 1,000 x g for 10 mins at 20-25°C and collect the serum fraction. Alternatively, a serum separator tube can be used following the manufacturer's instructions. Use immediately or store aliquots at -80°C. Avoid multiple freeze-thaw cycles.

Will ProcartaPlex multiplex assays give me the same results for each analyte as my current ELISA tests?

ProcartaPlex multiplex assays, which are based on Luminex xMAP technology, provide a versatile platform that gives users more flexibility and a greater array of options for analyte detection. Whether you are testing for single or multiple analytes, ProcartaPlex multiplex assays deliver accurate analytical performance using efficient, easy-to-follow protocols. Each of these assays has undergone the same development, validation, manufacturing, and quality control standardization we conduct for our ELISAs. Each lot of ProcartaPlex multiplex assays as well as ELISA assays is fully qualified with the appropriate sample type (i.e., species-specific serum, plasma, and cell culture supernatants), and each lot is evaluated based on the following performance characteristics:

Specificity-each analyte is screened to make sure there is no significant cross-reactivity with other analytes in the multiplex test
Sensitivity-each analyte is evaluated for both functional sensitivity (differentiation from background) and lower limit of detection (LLOD)
Precision/accuracy-multiplex assays have good intra-assay precision (<10% CV), inter-assay precision (<10% CV), and lot-to-lot consistency (<20% CV); these values are comparable to or better than most ELISA tests
ProcartaPlex multiplex assays are regularly tested against the matching ELISAs. Therefore, you can switch easily from ProcartaPlex assays to ELISA and vice versa with reliable results. Most of our ProcartaPlex assays use the same antibody pairs as our traditional plate-based ELISAs, resulting in high correlation (R2 > 0.9) between the two assays.

How does the Luminex xMAP technology work for measuring more than one analyte in the same well?

Luminex xMAP technology is based on polystyrene or paramagnetic microspheres, or beads, that are internally dyed with red and infrared fluorophores of differing intensities. Each dyed bead is given a unique number, known as a “bead region”, allowing the differentiation of beads. For ProcartaPlex multiplex immunoassay kits, individual bead sets are then coated with a capture antibody qualified for one specific analyte. Multiple analyte-specific beads can then be combined in a single well of a 96-well assay to detect and quantify multiple targets simultaneously, using one of the Luminex instruments for analysis.

What is the Luminex assay platform?

The Luminex assay is a bead-based immunoassay that uses beads of defined spectral properties conjugated to protein-specific capture antibodies and added along with samples (including standards of known protein concentration and test samples) into the wells of a microplate. The target protein binds to the capture antibodies over the course of a 2 hr incubation. After incubation on a shaker, the beads are washed by putting the 96-well plate on a flat magnet for 30 seconds, after which the fluid is discarded by flicking the wells or by using an automated plate washer. The magnet is removed, and the beads are resuspended in the detection antibody. Another incubation and wash are followed by the addition of streptavidin–R-phycoerythrin (SAPE). The beads are then washed and are ready to analyze. The Luminex technology is compatible with the following Luminex analyzers:

MAGPIX System-affordable, efficient, and compact
Luminex 200 System-versatile, efficient, and widely used in multiplexing
FLEXMAP 3D System-high throughput (up to 500 simultaneous assays) and automation compatible


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.