Learn More
Medchemexpress LLC HY-P7379 10ug Medchemexpress, IL-8/CXCL8 (CHO-expressed), Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP737910UG
Encompass_Preferred
Medchemexpress, HY-P7379 10ug IL-8/CXCL8 (CHO-expressed), Human CAS: Recombinant Human Interleukin-8/CXCL8 (CHO-expressed) is a proinflammatory CXC chemokine associated with the promotion of neutrophil chemotaxis and degranulation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3156