| Accession Number |
Q9XSX5 |
| Assay Range |
25-6000 pg/mL |
| Assay Sensitivity |
25 pg/mL |
| Calibration Range |
25 pg/mL to 6000 pg/mL |
| Assay |
Solid-phase sandwich ELISA |
| Conjugate |
HRP |
| Immunoassay Kit Format |
Sandwich ELISA Kit |
| Product Type |
ELISA |
| Protein Family |
GPCR (G-Protein Coupled Receptors) |
| Sample Type |
Plasma, Serum, Supernatant |
| Specificity |
This ELISA pair antibody detects feline IL-8. Other species not determined |
| Sufficient For |
96 Assays |
| Target Species |
Feline |
| For Use With (Application) |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis |
| For Use With (Equipment) |
Absorbance Microplate Reader |
| Gene Alias |
(Ala-IL-8)77; (Ser-IL-8)72; Alveolar macrophage chemotactic factor I; alveolar macrophage-derived chemotactic factor-I; AMCF-I; ANX7; beta endothelial cell-derived neutrophil activating peptide; beta-thromboglobulin-like protein; chemokine (C-X-C motif) ligand 8; C-X-C motif chemokine 8; C-X-C motif chemokine ligand 8; CXCL8; cytokine 8; emoctakin; GCP/IL-8 protein I; GCP/IL-8 protein II; GCP/IL-8 protein III; GCP/IL-8 protein IV; GCP/IL-8 protein V; GCP/IL-8 protein VI; GCP1; GCP-1; Granulocyte chemotactic protein 1; IL8; IL-8; IL-8(1-77); IL-8(5-77); IL-8(6-77); IL-8(7-77); IL-8(8-77); IL-8(9-77); IL8/NAP1 form I; IL8/NAP1 form II; IL8/NAP1 form III; IL8/NAP1 form IV; IL8/NAP1 form V; IL8/NAP1 form VI; ILN; Interleukin; interleukin 8; Interleukin8; interleukin-8; interleuklin-8 precursor; LECT; LUCT; lung giant cell carcinoma-derived chemotactic protein; lymphocyte derived neutrophil activating peptide; Lymphocyte-derived neutrophil-activating factor; LYNAP; MDNCF; MDNCF-a; MDNCF-b; MDNCF-c; MONAP; Monocyte-derived neutrophil chemotactic factor; monocyte-derived neutrophil-activating peptide; NAF; NAP1; NAP-1; neutrophil activating peptide 1; neutrophil and T-lymphocyte chemotactic and activating factor; Neutrophil attractant/activation protein 1; neutrophil attractant/activation protein-1; Neutrophil-activating factor; neutrophil-activating peptide 1; Neutrophil-activating protein 1; Permeability factor 1; PF1; Protein 3-10C; RP11-537A6.8; RPF1; small inducible cytokine subfamily B, member 8; SNX; SYNEXIN; T-cell chemotactic factor; tumor necrosis factor-induced gene 1(Ala-IL-8)77; (Ser-IL-8)72; Alveolar macrophage chemotactic factor I; alveolar macrophage-derived chemotactic factor-I; AMCF-I; ANX7; beta endothelial cell-derived neutrophil activating peptide; beta-thromboglobulin-like protein; chemokine (C-X-C motif) ligand 8; C-X-C motif chemokine 8; C-X-C motif chemokine ligand 8; CXCL8; cytokine 8; emoctakin; GCP/IL-8 protein I; GCP/IL-8 protein II; GCP/IL-8 protein III; GCP/IL-8 protein IV; GCP/IL-8 protein V; GCP/IL-8 protein VI; GCP1; GCP-1; Granulocyte chemotactic protein 1; IL8; IL-8; IL-8(1-77); IL-8(5-77); IL-8(6-77); IL-8(7-77); IL-8(8-77); IL-8(9-77); IL8/NAP1 form I; IL8/NAP1 form II; IL8/NAP1 form III; IL8/NAP1 form IV; IL8/NAP1 form V; IL8/NAP1 form VI; ILN; Interleukin; interleukin 8; Interleukin8; interleukin-8; interleuklin-8 precursor; LECT; LUCT; lung giant cell carcinoma-derived chemotactic protein; lymphocyte derived neutrophil activating peptide; Lymphocyte-derived neutrophil-activating factor; LYNAP; MDNCF; MDNCF-a; MDNCF-b; MDNCF-c; MONAP; Monocyte-derived neutrophil chemotactic factor; monocyte-derived neutrophil-activating peptide; NAF; NAP1; NAP-1; neutrophil activating peptide 1; neutrophil and T-lymphocyte chemotactic and activating factor; Neutrophil attractant/activation protein 1; neutrophil attractant/activation protein-1; Neutrophil-activating factor; neutrophil-activating peptide 1; Neutrophil-activating protein 1; Permeability factor 1; PF1; Protein 3-10C; RP11-537A6.8; RPF1; small inducible cytokine subfamily B, member 8; SNX; SYNEXIN; T-cell chemotactic factor; tumor necrosis factor-induced gene 1; IL8 |
| Gene ID (Entrez) |
493836 |
| Gene Symbol |
CXCL8 |
| Interassay CV |
<12% |
| Intraassay CV |
<10% |
| Kit Contents |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers |
| Label or Dye |
Biotin |
| Protein Aliases |
IL-8, C-X-C motif chemokine 8, Chemokine (C-X-C motif) ligand 8 |
| Quantity |
5 x 96 Tests |
| Regulatory Status |
RUO |
| Research Discipline |
Extracellular Matrix & Cell Adhesion |
| Sample Volume |
Plasma 50 μL, Serum 50 μL, Supernatant 100 μL |
| Shipping Condition |
Wet or Dry Ice |
| Storage Requirements |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C |
| Synonym |
IL8 |
| Target |
IL-8 |
| Test Time |
1 hr 20 min |
| Total Assay Time |
4 hr 45 min |
|
Show More
Show Less
|
|