Learn More
ACROBiosystems ACROBiosystems Human IL-8 / CXCL8 Protein, His Tag
Catalog No. 501956908
Encompass_Preferred
Recombinant Protein;1MG;Human IL-8, His Tag (IL8-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 28 - Ser 99 (Accession # P10145-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 10.3 kDa. The protein migrates as 13 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6908 Supplier ACROBiosystems Supplier No. IL8H52H31MG
Recombinant Protein;1MG;Human IL-8, His Tag (IL8-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 28 - Ser 99 (Accession # P10145-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 10.3 kDa. The protein migrates as 13 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.