Learn More
Enzo Life Sciences IL-8 (human), (recombinant) (25µg)
SDP
Catalog No. 502013972
Encompass_Preferred
Alternative name: Interleukin-8, CXCL8, Monocyte-derived neutrophil chemotactic factor, MDNCF, Neutrophil activating factor, NAF, NAP-1. Purity: ≥98% (SDS & HPLC). Formulation: Lyophilized with no additives. Source: Produced in E. coli. Long Term Storage: -20°C.
Catalog No. 50-201-3972 Supplier Enzo Life Sciences Supplier No. ENZPRT1850025
Alternative name: Interleukin-8, CXCL8, Monocyte-derived neutrophil chemotactic factor, MDNCF, Neutrophil activating factor, NAF, NAP-1. Purity: ≥98% (SDS & HPLC). Formulation: Lyophilized with no additives. Source: Produced in E. coli. Long Term Storage: -20°C.