Learn More
Enzo Life Sciences IL-8 monoclonal antibody (NYR-HIL8) (500µg)
SDP
Supplier:  Enzo Life Sciences ADI905110500
Encompass_Preferred
Clone: NYR-HIL8. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant human Interleukin-8. Species reactivity: Human. Applications: ELISA, IP, WB. Purity: Ion exchange purified. Formulation: Lyophilized. In PBS.
Catalog No. 50-200-6934