Learn More
Enzo Life Sciences IL-8 receptor, Type B (human) monoclonal antibody (HC 2) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804040C100
Encompass_Preferred
Clone: HC 2. Isotype: IgG1. Immunogen: Synthetic peptide (CXCR2-Amino terminus Met1-Tyr18) coupled to BSA. Species reactivity: Human. Applications: Flow Cytometry, IHC (FS), WB. Purity: Purified by Protein A chromatography. Formulation: Liquid. Purified antibody in TRIS/HCl containing 0.05% sodium azide.
Catalog No. 50-200-9756