Learn More
Genetex Inc GeneTex Anti-IL1 Beta Antibody, Rabbit Polyclonal, Purified By Antigen-Affinity Chromatography, 100 µg
SDP
Supplier:  Genetex Inc GTX74034
Encompass
The GeneTex Anti-IL1 beta antibody is a versatile rabbit polyclonal antibody that provides high specificity for IL-1 beta studies in various applications such as WB, ICC/IF, and ELISA. Designed for research into inflammatory responses, it selectively detects IL-1 beta across human, mouse, and rat samples. With its purified form by antigen-affinity chromatography, it ensures reliable results in your experimental contexts.
Catalog No. NC2257405