Learn More
Aviva Systems Biology Corp IL10RA Antibody - N-terminal region (ARP45135_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45135_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against IL10RA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human IL10RA. Alternate names: CDW210A, HIL-10R, IL-10R1, IL10R, CD210, CD210aApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rat
Catalog No. 50-014-58973