Learn More
Eurofins Discoverx PathHunter IL10RA/10RB Dimerization Bioassay Kit, 10pl. (93-0985Y3-00136)
Catalog No. 502001752
Encompass
The PathHunter IL10RA/IL10RB Dimerization Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses ligand (e.g. IL-10) induced dimerization of IL10RA/IL10RB receptors. This product uses EFC technology. The cell line included overexpresses IL10RA target protein fused to PK alongside IL10RB target protein fused to EA. Upon ligand-induced dimerization of IL10RA/IL10RB, detectable EFC signal is produced. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.
Catalog No. 50-200-1752 Supplier Eurofins Discoverx Supplier No. 930985Y300136
Add to Cart
Add to Cart
Request a Quote
The PathHunter IL10RA/IL10RB Dimerization Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses ligand (e.g. IL-10) induced dimerization of IL10RA/IL10RB receptors. This product uses EFC technology. The cell line included overexpresses IL10RA target protein fused to PK alongside IL10RB target protein fused to EA. Upon ligand-induced dimerization of IL10RA/IL10RB, detectable EFC signal is produced. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.