Learn More
Sino Biological IL-11 / interleukin 11 Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 12225MM09100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL-11 / interleukin 11 (rh IL-11 / interleukin 11; Catalog#12225-HNCE; AAH12506.1; Pro22-Leu199). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8664