Learn More
Biomatik Corporation IL11RA Polyclonal Antibody
Supplier:  Biomatik Corporation CAF50176
Encompass_Preferred
IL11RA Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Interleukin 11 Receptor, Alpha. The IL11RA Polyclonal Antibody has been validated for the following applications: ELISA, WB, IF, FC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Please Inquire. Applications: ELISA, WB, IF, FC. Target Synonyms: Interleukin 11 Receptor, Alpha. Accession Number: Q14626. Gene ID: 3590. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9225