Learn More
Sino Biological IL11RA Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 10252T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL11RA (rh IL11RA; Catalog#10252-H08H; NP_004503.1; Met1-Val363). IL11RA specific IgG was purified by Human IL11RA affinity chromatography.
Catalog No. 50-224-7444