Learn More
Sino Biological IL12 / IL12A & IL12B Antibody (Biotin), Rabbit MAb 100ug
Supplier:  Sino Biological CT011R070B100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL12 / IL12A & IL12B (rh IL12 / IL12A & IL12B; Catalog#CT011-H08H; P29459-1 & NP_002178.2; Met1-Ser219 & Met1-Ser328) and then biotinylated.
Catalog No. 50-224-6571