Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents QP520410UG
Encompass_Preferred
Product Type: Recombinant Protein. Conjugate/Tag: Untagged. Host: HEK 293. Species Reactivity: Mouse. BioActivity: BioActivity: Shows bioactivity at a concentration of 20 ng/mL in the Mouse primary T cell induced expression of CD25 assay. The bioactivity acceptance criteria is set at 5 x 10^4 units/mg. AA Sequence: See company website. Buffer: 10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5. Purity: Greater than 95% as determined by SDS-PAGE. Endotoxin: Less than or equal to 1.00 EUs/ug as determined by the LAL method. Gene Symbol: IL12. Gene ID: 16160. Uniprot: P43432; P43431.
Catalog No. 50-188-5707