Learn More
ACROBiosystems Recombinant Protein;Human IL-12A / NKSF1 / p35 Protein, Fc Tag;HEK293;500UG;NK1,IL12A,CLMF,IL-12A,NFSK,NKSF1,P35,NKSF-1,Interleukin
Catalog No. 501658742
Encompass_Preferred
Recombinant Protein;Human IL-12A / NKSF1 / p35 Protein, Fc Tag;500UG;Human IL-12A, Fc Tag (NK1-H5252) is expressed from human 293 cells (HEK293). It contains AA Arg 23 - Ser 219 (Accession # P29459).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 49.2 kDa. The protein migrates as 65-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8742 Supplier ACROBiosystems Supplier No. NK1H5252500UG
Recombinant Protein;Human IL-12A / NKSF1 / p35 Protein, Fc Tag;500UG;Human IL-12A, Fc Tag (NK1-H5252) is expressed from human 293 cells (HEK293). It contains AA Arg 23 - Ser 219 (Accession # P29459).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 49.2 kDa. The protein migrates as 65-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.