Learn More
Sino Biological IL13 / ALRH Antibody, Rabbit PAb 100uL
Supplier:  Sino Biological 11057RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Cynomolgus IL13 / ALRH (Catalog#11057-C07H; ABG75889.1; Ser21-Asn132). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-224-8173