Learn More
Aviva Systems Biology Corp IL13RA2 Antibody - N-terminal region (ARP53557_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53557_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IL13RA2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human IL13RA2. Alternate names: CD213A2, IL-13R, IL13BP, CT19ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61047