Learn More
ACROBiosystems Recombinant Protein;Mouse IL-13 R alpha 1 Protein, Fc Tag;HEK293;1MG;IL1,IL13RA1,CD213A1,IL-13Ra,NR4,RP13-128O4.2,IL13Rα1,Interleukin
Catalog No. 501658713
Encompass_Preferred
Recombinant Protein;Mouse IL-13 R alpha 1 Protein, Fc Tag;1MG;Mouse IL-13RA1, Fc Tag (IL1-M5258) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Thr 340 (Accession # NP_598751).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 62.7 kDa. The protein migrates as 67-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8713 Supplier ACROBiosystems Supplier No. IL1M52581MG
Recombinant Protein;Mouse IL-13 R alpha 1 Protein, Fc Tag;1MG;Mouse IL-13RA1, Fc Tag (IL1-M5258) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Thr 340 (Accession # NP_598751).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 62.7 kDa. The protein migrates as 67-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.