Learn More
ACROBiosystems Recombinant Protein;Mouse IL-13 R alpha 1 Protein;HEK293;100UG;IL1,IL13RA1,CD213A1,IL-13Ra,NR4,RP13-128O4.2,IL13Rα1,Interleukin
Catalog No. 501658714
Encompass_Preferred
Recombinant Protein;Mouse IL-13 R alpha 1 Protein;100UG;Mouse IL-13RA1, His Tag (IL1-M52H7) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Thr 340 (Accession # NP_598751).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.9 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8714 Supplier ACROBiosystems Supplier No. IL1M52H7100UG
Recombinant Protein;Mouse IL-13 R alpha 1 Protein;100UG;Mouse IL-13RA1, His Tag (IL1-M52H7) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Thr 340 (Accession # NP_598751).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.9 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.