Learn More
ACROBiosystems Recombinant Protein;Human IL-13 R alpha 1 Protein;HEK293;1MG;IL1,IL13RA1,CD213A1,IL-13Ra,NR4,RP13-128O4.2,IL13Rα1,Interleukin
Supplier:  ACROBiosystems IL1H52241MG
Encompass_Preferred
Recombinant Protein;Human IL-13 R alpha 1 Protein;1MG;Human IL-13RA1, His Tag (IL1-H5224) is expressed from human 293 cells (HEK293). It contains AA Gly 22 - Thr 343 (Accession # NP_001551.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 38 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-13RA1, His Tag (Cat. No. IL1-H5224) at 10μg/mL (100 μL/well) can bind Human IL-13, Fc Tag (Cat. No. IL3-H4269) with a linear range of 0.64-10 μg/mL (Routinely tested).
Catalog No. 50-165-7667