Learn More
Sino Biological Recombinant Human IL15 & IL15RA Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological CT094H02H100
Encompass_Preferred
A DNA sequence encoding the human IL15RA (NP_002180.1) (Ile31-Asp96) linked with the human IL15 (NP_000576.1, with mutation Asn 120 Asp) (Asn49-Ser162) by a peptide linker was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-211-5949