Learn More
Aviva Systems Biology Corp Il15ra Antibody - N-terminal region (ARP63493_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63493_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Il15ra. It was validated on Western Blot. . Alternate names: AA690181ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-987