Learn More
Sino Biological IL17 / IL17a Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 12047T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL17 / IL17a (rh IL17 / IL17a; Catalog#12047-HNAE; Q16552; Ile20-Ala155). IL17 / IL17a specific IgG was purified by Human IL17 / IL17a affinity chromatography.
Catalog No. 50-224-7707