Learn More
Sino Biological IL17 / IL17a Neutralizing Antibody 500ug
Supplier:  Sino Biological 12047M237500
Encompass_Preferred
This antibody was obtained from a mouse immunized with purified, recombinant Human IL17 / IL17a (rHuman IL17 / IL17a; Catalog#12047-HNAE; Q16552; Ile20-Ala155) and was produced using recombinant antibody technology.
Catalog No. 50-224-8258