Learn More
AdipoGen anti-IL-17A (human), mAb (17-142F) (50 µg)
SDP
Supplier:  AdipoGen AG20A0071C050
Encompass_Preferred
Monoclonal Antibody. Recognizes human IL-17A. Isotype: Mouse IgG1kappa. Clone: 17-142F. Applications: ELISA, FACS. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. IL-17A exhibits multiple biological activities on a variety of cells including the induction of IL-6 and IL-8 production in fibroblsts, the enhancement of surface expression of ICAM-1 in fibroblast, activation of NF-kappaB and costimulation of T cell proliferation. Induces stromal cells to produce proinflammatory and hematopoietic cytokines.
Catalog No. 50-205-2870