Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human IL-17A & IL-17F Protein, His Tag, Avi Tag (Avitag™);HEK293;200UG;ILF,IL-17A,Interleukin-17A,CTLA-8,IL-17,IL17A,Interleukin 17A,CTLA8,IL17,IL17F,IL-17F,IL-24,IL24,ML-1,ML1,Interleukin-24,Interleukin-17F,Interleukin 17F,Interleukin17F
Supplier:  ACROBiosystems ILFH82W1200UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human IL-17A & IL-17F Protein, His Tag, Avi Tag (Avitag™);200UG;Biotinylated Human IL-17A & IL-17F, His Tag, Avi Tag (ILF-H82W1) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Ala 155 (IL-17A) & Arg 31 - Gln 163 (IL-17F) (Accession # Q16552-1 (IL-17A) & Q96PD4-1 (IL-17F)).;Biotinylated Human IL-17A & IL-17F, His Tag, Avi Tag, produced by co-expression of IL-17A and IL-17F, has a calculated MW of 34.1 kDa. Subunit IL-17A & IL-17F is fused with an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The reducing (R) protein migrates as 40-43 kDa due to glycosylation.;Immobilized Anti-Human IL17A Mab at 2 μg/mL (100 μL/well) can bind Biotinylated Human IL-17A & IL-17F, His Tag, Avi Tag (Cat. No. ILF-H82W1) with a linear range of 0.2-6 ng/mL (QC tested).
Catalog No. 50-165-7546