Learn More
Aviva Systems Biology Corp IL17D Antibody - N-terminal region (ARP57724_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57724_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IL17D. It was validated on Western Blot. IL17D,interleukin 17D,IL27; IL-22; IL-27; IL-17D; FLJ30846; IL17D,interleukin-17D,interleukin-17D,interleukin 27,interleukin-27The immunogen is a synthetic peptide directed towards the N terminal region of human IL17D. Alternate names: FLJ30846, IL-17D, IL-22, IL-27, IL27ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse
Catalog No. 50-286-364