Learn More
Sino Biological IL17RA Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 51065R217100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL17RA (rM IL17RA; Catalog#51065-MNAE; Q62386; Thr22-Ala158).
Catalog No. 50-224-6536