Learn More
Sino Biological Human IL-17R / IL-17RA (CD217) Protein (Fc Tag) 100ug
Supplier:  Sino Biological 10895H02H100
Encompass_Preferred
A DNA sequence encoding the human IL17RA (NP_055154.3) (Met1-Trp320) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-211-4943