Learn More
ACROBiosystems Recombinant Protein;Human IL-17 RA / CD217 Protein;HEK293;1MG;ILA,CD217,CDw217,IL-17RA,CANDF5,IL17R,hIL-17R,IL17RA,Interleukin
Supplier:  ACROBiosystems ILAH52221MG
Encompass_Preferred
Recombinant Protein;Human IL-17 RA / CD217 Protein;1MG;Human IL-17 RA, His Tag (ILA-H5222) is expressed from human 293 cells (HEK293). It contains AA Leu 33 - Trp 320 (Accession # NP_055154.3).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 34.3 kDa. The protein migrates as 55-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-17A, Tag Free (Cat. No. ILA-H5118) at 0.5 μg/mL (100 μL/well) can bind Human IL-17 RA, His Tag (Cat. No. ILA-H5222) with a linear range of 4-31 ng/mL (QC tested).
Catalog No. 50-165-7827