Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human IL-17 RA / CD217 Protein, His Tag & Avi Tag (Avitag™);HEK293;25UG;ILR,CD217,CDw217,IL-17RA,CANDF5,IL17R,hIL-17R,IL17RA,Interleukin
Catalog No. 501658241
Encompass_Preferred
Recombinant Protein;Biotinylated Human IL-17 RA / CD217 Protein, His Tag & Avi Tag (Avitag™);25UG;Biotinylated Human IL-17 RA, His Tag, Avi Tag (ILR-H82E5) is expressed from human 293 cells (HEK293). It contains AA Leu 33 - Trp 320 (Accession # Q96F46-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 37.2 kDa. The protein migrates as 55-68 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-17A, Tag Free (Cat. No. ILA-H5118) at 2 μg/mL (100 μL/well) can bind Biotinylated Human IL-17 RA, His Tag, Avi Tag (Cat. No. ILR-H82E5) with a linear range of 0.2-8 ng/mL (QC tested).
Catalog No. 50-165-8241 Supplier ACROBiosystems Supplier No. ILRH82E525UG
Recombinant Protein;Biotinylated Human IL-17 RA / CD217 Protein, His Tag & Avi Tag (Avitag™);25UG;Biotinylated Human IL-17 RA, His Tag, Avi Tag (ILR-H82E5) is expressed from human 293 cells (HEK293). It contains AA Leu 33 - Trp 320 (Accession # Q96F46-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 37.2 kDa. The protein migrates as 55-68 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human IL-17A, Tag Free (Cat. No. ILA-H5118) at 2 μg/mL (100 μL/well) can bind Biotinylated Human IL-17 RA, His Tag, Avi Tag (Cat. No. ILR-H82E5) with a linear range of 0.2-8 ng/mL (QC tested).