Learn More
Aviva Systems Biology Corp IL18 Antibody - C-terminal region (ARP60740_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60740_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IL18. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human IL18. Alternate names: IGIF, IL-18, IL-1g, IL1F4, MGC12320ApplicationWB. Species reactivity: Human, Mouse, Rat
Catalog No. 50-287-145