Learn More
Sino Biological IL-18 Antibody (HRP), Rabbit MAb 100ug
Supplier:  Sino Biological 50073R167H100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL-18 (rM IL-18; Catalog#50073-MNAE; P70380; Asn36-Ser192), and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-224-7601