Learn More
Sino Biological IL-18 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 90011T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rhesus IL-18 ( Catalog#90011-C07E; NP_001028006.1; Ala2-Asp193). IL-18 specific IgG was purified by Rhesus IL-18 affinity chromatography.
Catalog No. 50-224-6553