Learn More
ABclonal Technology IL18 Rabbit PolymAb
SDP
Catalog No. 5000006752
Encompass_Preferred
by caspase-1 and is capable of stimulating interferon gamma production and of regulating both T helper (Th) 1 and Th2 responses This cytokine has been implicated in the injury of different organs and in potentially fatal conditions characterized by a cytokine storm In humans IL-18 gene is located on chromosome 11 Alternatively spliced transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-06752 Supplier ABclonal Technology Supplier No. A24169PM20UL
by caspase-1 and is capable of stimulating interferon gamma production and of regulating both T helper (Th) 1 and Th2 responses This cytokine has been implicated in the injury of different organs and in potentially fatal conditions characterized by a cytokine storm In humans IL-18 gene is located on chromosome 11 Alternatively spliced transcript variants encoding different isoforms have been found for this gene