Learn More
ACROBiosystems Recombinant Protein;Human IL-18BP Protein;HEK293;200UG;ILP,IL18BP,IL18BPa,Tadekinig-alfa,IL-18BP,IL-18BPa,IL-18 BP
Catalog No. 501658647
Encompass_Preferred
Recombinant Protein;Human IL-18BP Protein;200UG;Human IL-18BP, His Tag (ILP-H5222) is expressed from human 293 cells (HEK293). It contains AA Thr 31 - Gly 194 (Accession # NP_001138529).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.5 kDa. The protein migrates as 28-48 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8647 Supplier ACROBiosystems Supplier No. ILPH5222200UG
Recombinant Protein;Human IL-18BP Protein;200UG;Human IL-18BP, His Tag (ILP-H5222) is expressed from human 293 cells (HEK293). It contains AA Thr 31 - Gly 194 (Accession # NP_001138529).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 18.5 kDa. The protein migrates as 28-48 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.