Learn More
Aviva Systems Biology Corp IL18R1 Antibody - N-terminal region (ARP46382_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46382_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IL18R1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human IL18R1. Alternate names: CDw218a, IL-1Rrp, IL1RRP, CD218a, IL18RAApplicationWB. Species reactivity: Human
Catalog No. 50-014-59315