Learn More
R&D Systems™ IL18R1 Antibody Pair [HRP]

Description
Specifications
Specifications
| Assay Range | 6.25 pg/mL |
| Assay Sensitivity | 6.25-400 pg/mL |
| Assay | Sandwich ELISA |
| Conjugate | HRP |
| Product Line | Novus |
| Product Type | Antibody Pairs |
| Specificity | This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse IL18R1. |
| Target Species | Mouse |
| For Use With (Application) | Sandwich ELISA |
| Gene Alias | CD218 antigen-like family member A, CD218a, CD218a antigen, CDw218a, cytokine receptor, IL-18R1, IL-18R-1, IL1R-rp, IL1RRPIL1 receptor-related protein, IL-1RrpIL18RA, interleukin 18 receptor 1, interleukin-18 receptor 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.