Learn More
ACROBiosystems Recombinant Protein;Human IL-18 R1 / CD218a Protein, Fc Tag;HEK293;200UG;IL1,IL18R1,IL1RRP,CD218a,CDw218a,IL-1Rrp,IL1R-rp,IL-18R1,IL-1RRP,IL18Ra,IL-18Ra
Catalog No. 501658643
Encompass_Preferred
Recombinant Protein;Human IL-18 R1 / CD218a Protein, Fc Tag;200UG;Human IL-18 R1, Fc Tag (IL1-H5259) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - Arg 329 (Accession # AAH69575).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 62.4 kDa. The protein migrates as 90-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8643 Supplier ACROBiosystems Supplier No. IL1H5259200UG
Recombinant Protein;Human IL-18 R1 / CD218a Protein, Fc Tag;200UG;Human IL-18 R1, Fc Tag (IL1-H5259) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - Arg 329 (Accession # AAH69575).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 62.4 kDa. The protein migrates as 90-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.