Learn More
Antibodies Online IL18RAP antibody (AA 20-356) 100ug
SDP
Catalog No. NC3971036
Encompass
The Mouse Monoclonal anti-IL18RAP antibody is suitable to detect IL18RAP in samples from Human. It has been validated for WB and Neut.
Catalog No. NC3971036 Supplier Antibodies Online Supplier No. ABIN1983269
The Mouse Monoclonal anti-IL18RAP antibody is suitable to detect IL18RAP in samples from Human. It has been validated for WB and Neut.