Learn More
Aviva Systems Biology Corp IL1A Antibody - N-terminal region (ARP54322_P050) | 100uL
SDP
Catalog No. 5001461173
Encompass_Preferred
This is a rabbit polyclonal antibody against IL1A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human IL1A. Alternate names: IL-1A, IL1, IL1-ALPHA, IL1F1ApplicationWB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Rat, Sheep
Catalog No. 50-014-61173 Supplier Aviva Systems Biology Corp Supplier No. ARP54322_P050
This is a rabbit polyclonal antibody against IL1A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human IL1A. Alternate names: IL-1A, IL1, IL1-ALPHA, IL1F1ApplicationWB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Rat, Sheep