Learn More
Aviva Systems Biology Corp IL1B Antibody - N-terminal region (ARP54323_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54323_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IL1B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human IL1B. Alternate names: IL-1, IL1-BETA, IL1F2ApplicationIHC, WB. Species reactivity: Dog, Horse, Human, Mouse, Rabbit
Catalog No. 50-285-906