Learn More
Aviva Systems Biology Corp IL1F10 Antibody - middle region (ARP60267_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60267_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IL1F10. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human IL1F10. Alternate names: FIL1-theta, FKSG75, IL-1HY2, IL1-theta, MGC119831, MGC119832, MGC119833ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-286-990