Learn More
Aviva Systems Biology Corp Il1f5 Antibody - C-terminal region (ARP55627_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55627_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Il1f5. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Rat. Alternate names: Il1f5ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-062