Learn More
ACROBiosystems Recombinant Protein;Human IL-1 RAcP / IL-1 R3 Protein;HEK293;100UG;ILP,IL1RAP,IL-1RAcP,C3orf13,IL1R3,IL‑1 RAcP,IL‑1 R3,IL‑1R3,IL-1RAP,
Catalog No. 501658509
Encompass_Preferred
Recombinant Protein;Human IL-1 RAcP / IL-1 R3 Protein;100UG;Human IL-1 RAcP, His Tag (ILP-H5225) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Glu 359 (Accession # Q9NPH3).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 40 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-8509 Supplier ACROBiosystems Supplier No. ILPH5225100UG
Recombinant Protein;Human IL-1 RAcP / IL-1 R3 Protein;100UG;Human IL-1 RAcP, His Tag (ILP-H5225) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Glu 359 (Accession # Q9NPH3).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 40 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.